APOH0680BE4F Apollo Scientific Ltd

5-Bromo-2-chloropyridine-3-boronic acid

  • MDL number MFCD09037486
  • Cas Number : 1072944-19-4
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 148-150
Iupac Name 5-bromo-2-chloro-3-pyridinylboronic acid
Inchi Code 1S/C5H4BBrClNO2/c7-3-1-4(6(10)11)5(8)9-2-3/h1-2,10-11H
Inchi Key VDOTXMKSABQXLA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 148-150
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloropyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloropyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart