APOH0680BEA5 Apollo Scientific Ltd

3-Nitrophenylacetylene

  • MDL number MFCD02181150
  • Cas Number : 3034-94-4
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethynyl-3-nitrobenzene
Inchi Code 1S/C8H5NO2/c1-2-7-4-3-5-8(6-7)9(10)11/h1,3-6H
Inchi Key JOUOQPWPDONKKS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-nitrophenylacetylene

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-nitrophenylacetylene
pictograms GHS06
hstatements H301+H311+H331;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart