APOH0680BED9 Apollo Scientific Ltd

3-Bromo-2-iodobenzyl alcohol

  • Cas Number : 1261571-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-bromo-2-iodophenyl)methanol
Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=81898

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=81898
Availability details for
Quantity
  • Availability
  • Add to Cart