APOH0680C865 Apollo Scientific Ltd

4-Chloro-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline

  • MDL number MFCD01566524
  • Cas Number : 503148-24-1
  • Molecular Weight : 315.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-6-trifluoromethoxy-2-trifluoromethylquinoline

Safety Information

MSDS https://store.apolloscientific.co.uk/product/4-chloro-6-trifluoromethoxy-2-trifluoromethylquinoline
Availability details for
Quantity
  • Availability
  • Add to Cart