APOH0680D47B Apollo Scientific Ltd

4,4-Bis(trifluoromethyl)-5,5,6,6,7,7,7-heptafluoroheptyl acrylate

  • MDL number MFCD28334481
  • Cas Number : 1980085-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/44-bistrifluoromethyl-5566777-heptafluoroheptyl-acrylate

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/44-bistrifluoromethyl-5566777-heptafluoroheptyl-acrylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart