APOH086A1EC2 Apollo Scientific Ltd

(3R)-3-Methylmorpholine hydrochloride

  • MDL number MFCD18382515
  • Cas Number : 953780-78-4
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3r-3-methylmorpholine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3r-3-methylmorpholine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart