APOH086A2025 Apollo Scientific Ltd

6-Amino-5-chloronicotinonitrile

  • MDL number MFCD03617768
  • Cas Number : 156361-02-3
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 188-198
Iupac Name 6-amino-5-chloronicotinonitrile
Inchi Code 1S/C6H4ClN3/c7-5-1-4(2-8)3-10-6(5)9/h1,3H,(H2,9,10)
Inchi Key XDDIMILABMXHFG-UHFFFAOYSA-N

Description

Synonym 6-Amino-5-chloropyridine-3-carbonitrile, 2-Amino-3-chloro-5-cyanopyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 188-198
Product Link https://store.apolloscientific.co.uk/product/6-amino-5-chloronicotinonitrile

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/6-amino-5-chloronicotinonitrile
pictograms GHS06;GHS07;GHS05
hstatements H301;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart