APOH086A506D Apollo Scientific Ltd

3-Bromo-5-isopropylbenzoic acid

  • MDL number MFCD20040502
  • Cas Number : 112930-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-isopropylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-isopropylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart