APOH086A557E Apollo Scientific Ltd

2-Bromo-3-methoxybenzonitrile

  • MDL number MFCD09834772
  • Cas Number : 1261816-95-8
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3-methoxybenzonitrile
Inchi Code 1S/C8H6BrNO/c1-11-7-4-2-3-6(5-10)8(7)9/h2-4H,1H3
Inchi Key FQKUIMZTAJYITH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-methoxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-methoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart