APOH086A6BBA Apollo Scientific Ltd

4-Bromo-5-chloro-2-nitroaniline

  • MDL number MFCD11100159
  • Cas Number : 827-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-200
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-2-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-5-chloro-2-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart