APOH086A6D11 Apollo Scientific Ltd

Fmoc-L-HomoArg(Pbf)-OH

  • MDL number MFCD03093571
  • Cas Number : 1159680-21-3
  • Molecular Weight : 662.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/fmoc-l-homoargpbf-oh

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/fmoc-l-homoargpbf-oh
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart