APOH086A6DDE Apollo Scientific Ltd

N-Formylsaccharin

  • MDL number MFCD23380208
  • Cas Number : 50978-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1,3-Trioxo-1,2-benzothiazole-2-carbaldehyde
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=116203

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=116203
Availability details for
Quantity
  • Availability
  • Add to Cart