APOH086AAE6E Apollo Scientific Ltd

2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine

  • MDL number MFCD12024303
  • Cas Number : 887707-23-5
  • Molecular Weight : 289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-iodo-3-(trifluoromethyl)-2-pyridinol
Inchi Code 1S/C6H3F3INO/c7-6(8,9)4-1-3(10)2-11-5(4)12/h1-2H,(H,11,12)
Inchi Key ACMILMILHHLANU-UHFFFAOYSA-N

Description

Synonym 5-Iodo-3-(trifluoromethyl)pyridin-2-ol
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-5-iodo-3-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-5-iodo-3-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart