APOH086AAE70 Apollo Scientific Ltd

2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD31580162
  • Cas Number : 2090690-37-0
  • Molecular Weight : 273.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl]methanol
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-3-trifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-3-trifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart