APOH086AAE87 Apollo Scientific Ltd

2-bromo-6-fluoro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD30497608
  • Cas Number : 2059937-52-7
  • Molecular Weight : 287.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-6-fluoro-3-(trifluoromethyl)benzoic acid
Inchi Code 1S/C8H3BrF4O2/c9-6-3(8(11,12)13)1-2-4(10)5(6)7(14)15/h1-2H,(H,14,15)
Inchi Key XVUFMURDJMWSCI-UHFFFAOYSA-N

Description

Synonym 2-bromo-6-fluoro-3-(trifluoromethyl)benzoic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoro-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart