APOH086AC5DF Apollo Scientific Ltd

(2S,4S)-Fmoc-4-trifluoromethylpyrrolidine-2-carboxylic acid

  • MDL number MFCD11226824
  • Cas Number : 1242934-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=115633

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=115633
Availability details for
Quantity
  • Availability
  • Add to Cart