APOH086AC5E0 Apollo Scientific Ltd

5-Bromo-7-fluorobenzofuran

  • MDL number MFCD09056787
  • Cas Number : 286836-04-2
  • Molecular Weight : 215.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-7-fluoro-1-benzofuran
Inchi Code 1S/C8H4BrFO/c9-6-3-5-1-2-11-8(5)7(10)4-6/h1-4H
Inchi Key KKVACMAATHFELM-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-7-fluorobenzofuran

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/5-bromo-7-fluorobenzofuran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart