APOH086AC5E3 Apollo Scientific Ltd

2-Fluoro-3-methylbenzyl alcohol

  • MDL number MFCD07368780
  • Cas Number : 307975-03-7
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart