APOH0D1FFFB6 Apollo Scientific Ltd

(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride

  • MDL number MFCD31621177
  • Cas Number : 2231676-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=117614

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=117614
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart