APOH0D1FFFD6 Apollo Scientific Ltd

4-Bromo-1H-indole-6-carboxylic acid

  • MDL number MFCD03095205
  • Cas Number : 374633-27-9
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1h-indole-6-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1h-indole-6-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart