APOH0D1FFFED Apollo Scientific Ltd

1-Chloro-2,7-naphthyridine

  • MDL number MFCD00234389
  • Cas Number : 69042-30-4
  • Molecular Weight : 164.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-chloro-27-naphthyridine

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/1-chloro-27-naphthyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart