APOH0D200030 Apollo Scientific Ltd

5-Bromo-2-(trifluoromethyl)isonicotinonitrile

  • MDL number MFCD18261445
  • Cas Number : 1070892-04-4
  • Molecular Weight : 251.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-(trifluoromethyl)pyridine-4-carbonitrile
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethylisonicotinonitrile

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethylisonicotinonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart