APOH0D200051 Apollo Scientific Ltd

2-Bromo-3-fluoro-5-methylaniline

  • MDL number MFCD28739689
  • Cas Number : 1805421-04-8
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-3-fluoro-5-methylaniline
Inchi Code 1S/C7H7BrFN/c1-4-2-5(9)7(8)6(10)3-4/h2-3H,10H2,1H3
Inchi Key PNSMOWSOJAKJGX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoro-5-methylaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluoro-5-methylaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart