APOH0D2000A9 Apollo Scientific Ltd

1-(6-Chloro-5-fluoropyridin-3-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD18254428
  • Cas Number : 1245915-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=117503

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=117503
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart