APOH0D2000B0 Apollo Scientific Ltd

2-(Difluoromethoxy)pyridine-3-carbonitrile

  • MDL number MFCD14698406
  • Cas Number : 1214388-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=117513

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=117513
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart