APOH0D2000D3 Apollo Scientific Ltd

3-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid

  • MDL number MFCD28751019
  • Cas Number : 1805955-56-9
  • Molecular Weight : 223.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-difluoromethoxypyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-difluoromethoxypyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart