APOH0D2000E5 Apollo Scientific Ltd

4-(Difluoromethoxy)pyridine-2-carbaldehyde

  • MDL number MFCD22552055
  • Cas Number : 1315361-42-2
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(difluoromethoxy)picolinaldehyde
Inchi Code 1S/C7H5F2NO2/c8-7(9)12-6-1-2-10-5(3-6)4-11/h1-4,7H
Inchi Key HVFGATFZQSTWAA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxypyridine-2-carbaldehyde

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxypyridine-2-carbaldehyde
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart