APOH0D200110 Apollo Scientific Ltd

7-Fluoroimidazo[1,2-a]pyridine

  • MDL number MFCD11976201
  • Cas Number : 1260903-17-0
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-fluoroimidazo[1,2-a]pyridine
Inchi Code 1S/C7H5FN2/c8-6-1-3-10-4-2-9-7(10)5-6/h1-5H
Inchi Key DVDXNVNBHVEIOD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-fluoroimidazo12-apyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-fluoroimidazo12-apyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart