APOH0D200147 Apollo Scientific Ltd

5-Methyl-2-(trifluoromethyl)aniline

  • MDL number MFCD04972928
  • Cas Number : 106877-29-6
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-2-(trifluoromethyl)phenylamine
Inchi Code 1S/C8H8F3N/c1-5-2-3-6(7(12)4-5)8(9,10)11/h2-4H,12H2,1H3
Inchi Key HYQKJEMNTMFLJA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-2-trifluoromethylaniline

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-methyl-2-trifluoromethylaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart