APOH0D200209 Apollo Scientific Ltd

6-Amino-3-bromo-2-fluorobenzonitrile

  • MDL number MFCD06657979
  • Cas Number : 845866-92-4
  • Molecular Weight : 215.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-3-bromo-2-fluorobenzonitrile
Inchi Code 1S/C7H4BrFN2/c8-5-1-2-6(11)4(3-10)7(5)9/h1-2H,11H2
Inchi Key GGJQZVKWAKHTLK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-amino-3-bromo-2-fluorobenzonitrile

Safety Information

signalword Warning
pcodes P260;P301+P312;P280
MSDS https://store.apolloscientific.co.uk/product/6-amino-3-bromo-2-fluorobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart