APOH0D200215 Apollo Scientific Ltd

2,3-Difluoro-4-iodopyridine

  • MDL number MFCD09909609
  • Cas Number : 851386-34-0
  • Molecular Weight : 240.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 61-63
Iupac Name 2,3-difluoro-4-iodopyridine
Inchi Code 1S/C5H2F2IN/c6-4-3(8)1-2-9-5(4)7/h1-2H
Inchi Key AIXBVJWFQNYGJA-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 61-63
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-iodopyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-iodopyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart