APOH0D73E884 Apollo Scientific Ltd

3-Chloro-2-methyl-6-nitroaniline

  • MDL number MFCD09026962
  • Cas Number : 51123-59-2
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2-methyl-6-nitroaniline
Inchi Code 1S/C7H7ClN2O2/c1-4-5(8)2-3-6(7(4)9)10(11)12/h2-3H,9H2,1H3
Inchi Key ZFPIXTGBHCYNRN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-methyl-6-nitroaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-methyl-6-nitroaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart