APOH0D73E88A Apollo Scientific Ltd

3-(3-Pyridyl)-D-alanine hydrochloride

  • MDL number MFCD03095567
  • Cas Number : 350228-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym H-3-D-Pal-OH.HCl, (2R)-2-amino-3-pyridin-3-ylpropanoic acid hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-3-pyridyl-d-alanine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-3-pyridyl-d-alanine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart