APOH0D73E894 Apollo Scientific Ltd

Disodium 4-formylbenzene-1,3-disulfonate

  • MDL number MFCD00043913
  • Cas Number : 33513-44-9
  • Molecular Weight : 310.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Disodium 4-formylbenzene-1,3-disulfonate
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/disodium-4-formylbenzene-13-disulfonate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/disodium-4-formylbenzene-13-disulfonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart