APOH0D73E89C Apollo Scientific Ltd

2-Hydroxy-6-(trifluoromethyl)benzaldehyde

  • MDL number MFCD16999701
  • Cas Number : 58914-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-3-(trifluoromethyl)phenol, 2-Formyl-3-hydroxybenzotrifluoride, 6-(Trifluoromethyl)salicylaldehyde
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49315

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49315
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart