APOH0D73E8A2 Apollo Scientific Ltd

2,3-Dibromo-1,4-difluorobenzene

  • MDL number MFCD12922620
  • Cas Number : 179737-33-8
  • Molecular Weight : 271.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dibromo-1,4-difluorobenzene
Inchi Code 1S/C6H2Br2F2/c7-5-3(9)1-2-4(10)6(5)8/h1-2H
Inchi Key TXTZEQOJTJIFAH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dibromo-14-difluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-dibromo-14-difluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart