APOH10B58F52 Apollo Scientific Ltd

3,6-Dichloroimidazo[1,2-a]pyridine-2-carbohydrazide

  • MDL number MFCD31629938
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-dichloroimidazo12-apyridine-2-carbohydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/36-dichloroimidazo12-apyridine-2-carbohydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart