APOH10B58FFF Apollo Scientific Ltd

6-Chloroimidazo[1,2-a]pyridine-3-carbohydrazide

  • MDL number MFCD06496218
  • Cas Number : 900018-73-7
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloroimidazo12-apyridine-3-carbohydrazide

Safety Information

signalword Warning
pcodes P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/6-chloroimidazo12-apyridine-3-carbohydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart