APOH10B59012 Apollo Scientific Ltd

2,4-Dichloro-N-methylbenzenesulfonamide 1g pack

  • MDL number MFCD03989572
  • Cas Number : 565181-75-1
  • Molecular Weight : 240.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-n-methylbenzenesulfonamide

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart