APOH10B5901A Apollo Scientific Ltd

2-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine

  • MDL number MFCD18427849
  • Cas Number : 26323-62-6
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-34-dihydro-2h-pyrido32-b14oxazine

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-methyl-34-dihydro-2h-pyrido32-b14oxazine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart