APOH11A9BBCE Apollo Scientific Ltd

Tris(2-phenylpyridine)iridium

  • MDL number MFCD04039766
  • Cas Number : 94928-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/tris2-phenylpyridineiridium

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/tris2-phenylpyridineiridium
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart