APOH11A9BBD1 Apollo Scientific Ltd

4-Bromo-3-chloro-2-fluorobenzaldehyde

  • MDL number MFCD26958726
  • Cas Number : 1696224-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-79
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluorobenzaldehyde-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluorobenzaldehyde-97
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart