APOH11A9BFA5 Apollo Scientific Ltd

3-Fluoro-4-hydroxy-L-phenylalanine

  • MDL number MFCD00002607
  • Cas Number : 7423-96-3
  • Molecular Weight : 199.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-261
Synonym (2S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid, 3-Fluoro-L-tyrosine
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxy-l-phenylalanine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxy-l-phenylalanine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart