APOH11A9C000 Apollo Scientific Ltd

Methyl 2-chloro-4-(trifluoromethyl)cinnamate

  • MDL number MFCD01313366
  • Cas Number : 243977-24-4
  • Molecular Weight : 264.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-[2-chloro-4-(trifluoromethyl)phenyl]acrylate, Methyl 3-[2-chloro-4-(trifluoromethyl)phenyl]prop-2-enoate
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloro-4-trifluoromethylcinnamate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloro-4-trifluoromethylcinnamate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart