APOH11A9C06C Apollo Scientific Ltd

2-(Perfluoro-5-methylhexyl)ethyl methacrylate 5g pack

  • MDL number MFCD00236098
  • Cas Number : 50836-66-3
  • Molecular Weight : 482.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H,2H,2H-Perfluoro(7-methyloctyl)methacrylate
Purity 97
Density 1.567
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-perfluoro-5-methylhexylethyl-methacrylate

Safety Information

signalword Warning
pcodes P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart