APOH11A9C0F1 Apollo Scientific Ltd

6-(2,2,2-Trifluoroethoxy)thionicotinamide

  • MDL number MFCD00277205
  • Cas Number : 175277-59-5
  • Molecular Weight : 236.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-123
Synonym 6-(2,2,2-Trifluoroethoxy)pyridine-3-thiocarboxamide, 6-(2,2,2-Trifluoroethoxy)pyridine-3-carbothioamide
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-222-trifluoroethoxythionicotinamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-222-trifluoroethoxythionicotinamide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart