APOH11A9C110 Apollo Scientific Ltd

3-(Trifluoromethyl)cinnamic acid

  • MDL number MFCD00004393
  • Cas Number : 779-89-5
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-136
Synonym 3-[3-(Trifluoromethyl)phenyl]acrylic acid, 3-[3-(Trifluoromethyl)phenyl]prop-2-enoic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethylcinnamic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethylcinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart