APOH11A9C11C Apollo Scientific Ltd

4-[4-(Trifluoromethyl)phenyl]but-3-en-2-one

  • MDL number MFCD00052848
  • Cas Number : 80992-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [4-(Trifluoromethyl)benzylidene]acetone, 4-(3-Oxobut-1-en-1-yl)benzotrifluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 118/4mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5391

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5391
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart