APOH11A9C1DC Apollo Scientific Ltd

2-Bromo-1,3,3,3-tetrafluoroprop-1-ene

  • MDL number MFCD03094566
  • Cas Number : 396716-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-1-(trifluoromethyl)ethene
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 29-32
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6145

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6145
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart