APOH11A9C201 Apollo Scientific Ltd

4'-Fluoro-2-hydroxychalcone

  • MDL number MFCD03788512
  • Cas Number : 76626-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Fluorophenyl)-3-(2-hydroxyphenyl)prop-2-en-1-one, 2-[3-(4-Fluorophenyl)-3-oxoprop-1-en-1-yl)phenol
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6335

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6335
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart